3,4-Dimethylbenzylalkohol

Lieferant: Thermo Scientific

Synonyme: 4-(Hydroxymethyl)-o-xylene, (3,4-Dimethylphenyl)methanol

TL00645.1 TL00645.10 TL00645.50
MAYBTL00645.1EA 0 EUR
MAYBTL00645.1 MAYBTL00645.10 MAYBTL00645.50
3,4-Dimethylbenzylalkohol
3,4-Dimethylbenzylalkohol
Formel: C₉H₁₂O
Molecular Weight: 136.19 g/mol
Siedepunkt: 218
Schmelzpunkt: 63
MDL: MFCD00004647
CAS-Nummer: 6966-10-5
EINECS: 230-175-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR