1-Brom-3-chlor-4-iodbenzol

Lieferant: Thermo Scientific

Synonyme: 4-Brom-2-chlor-1-iodbenzol

TL00909.250 TL00909.1
MAYBTL00909.250EA 0 EUR
MAYBTL00909.250 MAYBTL00909.1
1-Brom-3-chlor-4-iodbenzol
4-Bromo-2-chloro-1-iodobenzene
Formel: C₆H₃BrClI
Molecular Weight: 317,35 g/mol
MDL: MFCD00079709
CAS-Nummer: 31928-47-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR