2-Amino-5-chlorbenzoxazol

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-5-chlor-1,3-benzoxazol, Zoxazolamin

001002-1G 001002-5G 001002-25G
FLUO001002-1GEA 69.8 EUR
FLUO001002-1G FLUO001002-5G FLUO001002-25G
2-Amino-5-chlorbenzoxazol
2-Amino-5-chlorobenzoxazole
Formel: C₇H₅ClN₂O
Molecular Weight: 168,58 g/mol
Siedepunkt: 309 °C (1013 hPa)
Schmelzpunkt: 184 °C
MDL: MFCD00005770
CAS-Nummer: 61-80-3
EINECS: 200-519-4
Merck Index: 12,10328

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR