2-Iodcumol

Lieferant: FLUOROCHEM

Synonyme: 2-Iodisopropylbenzol, 2-(2-Iodphenyl)propan

001198.5G 001198.25G
FLUO001198.5GEA 53.6 EUR
FLUO001198.5G FLUO001198.25G
2-Iodcumol
2-Iodcumol
Formel: C₉H₁₁I
Molecular Weight: 246,09 g/mol
MDL: MFCD00013704
CAS-Nummer: 19099-54-8
EINECS: 242-819-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR