1-Brom-3-phenoxypropan

Lieferant: FLUOROCHEM

Synonyme: 3-Phenoxypropylbromid, 3-Brompropylphenylether

001213-100G 001213-25G
FLUO001213-100GEA 155 EUR
FLUO001213-100G FLUO001213-25G
1-Brom-3-phenoxypropan
1-Brom-3-phenoxypropan
Formel: (C₆H₅)OCH₂CH₂CH₂Br
Molecular Weight: 215,09 g/mol
Siedepunkt: 262 °C (1013 hPa)
Dichte: 1,365 g/cm³ (16 °C)
MDL: MFCD00000256
CAS-Nummer: 588-63-6
EINECS: 209-623-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR