2-Mercaptobenzylalkohol
Lieferant: FLUOROCHEM
001239.1G
001239-5G
001239-25G
FLUO001239.1GEA
41
EUR
FLUO001239.1G
FLUO001239-5G
FLUO001239-25G
2-Mercaptobenzylalkohol
2-Mercaptobenzylalkohol
Formel:
C₇H₈OS Molecular Weight: 140,21 g/mol Siedepunkt: 85 °C (0,001 mmHg) Schmelzpunkt: 33 °C Dichte: 1,21 g/cm³ (20 °C) |
MDL:
MFCD00014448 CAS-Nummer: 4521-31-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...