3,3,3-Trifluor-1-propanol

Lieferant: FLUOROCHEM

Synonyme: 3,3,3-Trifluoropropan-1-ol

001557-1G 001557-5G
FLUO001557-1GEA 42.9 EUR
FLUO001557-1G FLUO001557-5G
3,3,3-Trifluor-1-propanol
3,3,3-Trifluor-1-propanol
Formel: C₃H₅F₃O
Molecular Weight: 114.07 g/mol
Siedepunkt: 96…98 °C (1013 hPa)
Dichte: 1.3739 g/cm³ (25 °C)
MDL: MFCD00190649
CAS-Nummer: 2240-88-2
UN: 1987
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR