3-Fluor-p-toluonitril
Lieferant: FLUOROCHEM
Synonyme:
3-Fluoro-4-methylbenzonitrile
002156-25G
002156-100G
FLUO002156-25GEA
57.9
EUR
FLUO002156-25G
FLUO002156-100G
3-Fluor-p-toluonitril
3-Fluor-p-toluonitril
Formel:
C₈H₆FN Molecular Weight: 135,14 g/mol |
MDL:
MFCD00153174 CAS-Nummer: 170572-49-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...