2,2,3,3-Tetrafluor-1,4-butandiol
Lieferant: FLUOROCHEM
Synonyme:
2,2,3,3-Tetrafluorobutane-1,4-diol
002302-5G
002302-25G
FLUO002302-5GEA
94.1
EUR
FLUO002302-5G
FLUO002302-25G
2,2,3,3-Tetrafluor-1,4-butandiol
2,2,3,3-Tetrafluoro-1,4-butanediol
Formel:
C₄H₆F₄O₂ Molecular Weight: 162,08 g/mol Siedepunkt: 110…112 °C (13 mmHg) Schmelzpunkt: 85 °C |
MDL:
MFCD00042375 CAS-Nummer: 425-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...