3-(4-Aminophenyl)propionsäure
Lieferant: FLUOROCHEM
Synonyme:
3-(4-Aminophenyl)propansäure
002500-5G
002500-25G
FLUO002500-5GEA
189
EUR
FLUO002500-5G
FLUO002500-25G
3-(4-Aminophenyl)propionsäure
3-(4-Aminophenyl)propionsäure
Formel:
C₉H₁₁NO₂ |
MDL:
MFCD00017118 CAS-Nummer: 2393-17-1 EINECS: 219-245-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...