4-Isopropylphenylisothiocyanat
Lieferant: FLUOROCHEM
002627-5G
002627-1G
002627-25G
FLUO002627-5GEA
0
EUR
FLUO002627-5G
FLUO002627-1G
FLUO002627-25G
4-Isopropylphenylisothiocyanat
4-Isopropylphenylisothiocyanat
Formel:
C₁₀H₁₁NS Molecular Weight: 177,27 g/mol Siedepunkt: 252 °C (1013 hPa) Dichte: 1,03 g/cm³ (20 °C) |
MDL:
MFCD00022060 CAS-Nummer: 89007-45-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...