N-(3-Brompropyl)phthalimid

Lieferant: FLUOROCHEM

Synonyme: N-(3-Brompropyl)phthalsäureimid, N-(3-Brompropyl)-1,2-benzoldicarbonsäureimid

002710-25G 002710-100G
FLUO002710-25GEA 42.9 EUR
FLUO002710-25G FLUO002710-100G
N-(3-Brompropyl)phthalimid
N-(3-Brompropyl)phthalimid
Formel: C₁₁H₁₀BrNO₂
Molecular Weight: 268,11 g/mol
Siedepunkt: 433 °C (1013 hPa)
Schmelzpunkt: 70…73 °C
MDL: MFCD00005904
CAS-Nummer: 5460-29-7
EINECS: 226-738-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR