4,6-Dihydroxypyrimidin

Lieferant: FLUOROCHEM

Synonyme: 6-Hydroxy-1H-pyrimidin-4-on

002991-100G 002991.500G 002991-25G
FLUO002991-100GEA 134 EUR
FLUO002991-100G FLUO002991.500G FLUO002991-25G
4,6-Dihydroxypyrimidin
4,6-Dihydroxypyrimidine
Formel: C₄H₄N₂O₂
Schmelzpunkt: > 300 °C
MDL: MFCD00016733
CAS-Nummer: 1193-24-4
EINECS: 214-772-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR