2-Thenoesäure

Lieferant: FLUOROCHEM

Synonyme: Thiophen-2-carbonsäure, 2-Thiophencarbonsäure

003042-100G 003042-500G
FLUO003042-100GEA 104 EUR
FLUO003042-100G FLUO003042-500G
2-Thenoesäure
2-Thenoesäure
Formel: C₅H₄O₂S
Molecular Weight: 128,15 g/mol
Siedepunkt: 260 °C (1013 hPa)
Schmelzpunkt: 123…125 °C
MDL: MFCD00005437
CAS-Nummer: 527-72-0
EINECS: 208-423-4
Merck Index: 12,09491

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR