Tetrafluorisophthalonitril
Lieferant: FLUOROCHEM
Synonyme:
Tetrafluorbenzol-1,3-dicarbonitril, Tetrafluorisophthalsäuredinitril
003317-5G
003317-25G
FLUO003317-5GEA
209
EUR
FLUO003317-5G
FLUO003317-25G
Tetrafluorisophthalonitril
Tetrafluorisophthalonitril
Formel:
C₈F₄N₂ Molecular Weight: 200,1 g/mol Siedepunkt: 283 °C (1013 hPa) Schmelzpunkt: 76…78 °C |
MDL:
MFCD00013290 CAS-Nummer: 2377-81-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...