5-(Trifluormethyl)uracil

Lieferant: FLUOROCHEM

Synonyme: 2,4-Dihydroxy-5-(trifluormethyl)pyrimidin

003333-1G 003333-5G
FLUO003333-1GEA 30.8 EUR
FLUO003333-1G FLUO003333-5G
5-(Trifluormethyl)uracil
5-(Trifluormethyl)uracil
Formel: C₅H₃F₃N₂O₂
Molecular Weight: 180.09 g/mol
Siedepunkt: 350
Schmelzpunkt: 251…252 °C
MDL: MFCD00006024
CAS-Nummer: 54-20-6
EINECS: 200-197-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR