3-Thenoesäure

Lieferant: FLUOROCHEM

Synonyme: Thiophen-3-carbonsäure, 3-Thiophencarbonsäure

003460-5G 003460-100G 003460-25G
FLUO003460-5GEA 0 EUR
FLUO003460-5G FLUO003460-100G FLUO003460-25G
3-Thenoesäure
3-Thenoesäure
Formel: C₅H₄O₂S
Molecular Weight: 128,15 g/mol
Schmelzpunkt: 137 °C
MDL: MFCD00005467
CAS-Nummer: 88-13-1
EINECS: 201-802-5
Merck Index: 12,09415

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR