4-tert-Butylbenzolsulfonylchlorid
Lieferant: FLUOROCHEM
CFF859.
003698-25G
003698-100G
003698-10G
CHMPCFF859.EA
0
EUR
CHMPCFF859.
FLUO003698-25G
FLUO003698-100G
FLUO003698-10G
4-tert-Butylbenzolsulfonylchlorid
4-tert-Butylbenzolsulfonylchlorid
Formel:
C₁₀H₁₃ClO₂S Siedepunkt: 165 °C (18 mmHg) Schmelzpunkt: 83 °C |
MDL:
MFCD00007449 CAS-Nummer: 15084-51-2 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...