3-Brom-4,5-diaminobenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 3-Brom-5-(trifluormethyl)benzol-1,2-diamin, 1-Brom-2,3-diamino-5-(trifluormethyl)benzol, 3-Brom-5-(trifluormethyl)-1,2-phenylendiamin, 3-Brom-5-(trifluormethyl)-o-phenylendiamin, 3-Brom-4,5-diamino-α,α,α-trifluortoluol

004175-1G 004175-5G
FLUO004175-1GEA 36.9 EUR
FLUO004175-1G FLUO004175-5G
3-Brom-4,5-diaminobenzotrifluorid
3-Brom-4,5-diaminobenzotrifluorid
Formel: C₇H₆BrF₃N₂
MDL: MFCD00042484
CAS-Nummer: 113170-72-2
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR