4-Brom-α,α,α,2,3,5,6-heptafluortoluol

Lieferant: FLUOROCHEM

Synonyme: 4-Bromo-2,3,5,6-tetrafluorobenzotrifluoride, 1-Bromo-2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene

004275-25G 004275-100G
FLUO004275-25GEA 0 EUR
FLUO004275-25G FLUO004275-100G
4-Brom-α,α,α,2,3,5,6-heptafluortoluol
4-Brom-α,α,α,2,3,5,6-heptafluortoluol
Formel: C₇BrF₇
Molecular Weight: 296,97 g/mol
Siedepunkt: 151…153 °C (1013 hPa)
Dichte: 1,92 g/cm³ (20 °C)
MDL: MFCD00075240
CAS-Nummer: 17823-46-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR