3-Chlor-4-fluorbenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 1-Chloro-2-fluoro-5-(trifluoromethyl)benzene, 3-Chloro-4-fluorobenzotrifluoride, 2-Chloro-1-fluoro-4-(trifluoromethyl)benzene

004437-5G 004437-10G 004437-1G
FLUO004437-5GEA 31.1 EUR
FLUO004437-5G FLUO004437-10G FLUO004437-1G
3-Chlor-4-fluorbenzotrifluorid
3-Chlor-4-fluorbenzotrifluorid
Formel: C₇H₃ClF₄
MDL: MFCD00042207
CAS-Nummer: 78068-85-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR