2-Chlor-6-fluorbenzylamin

Lieferant: FLUOROCHEM

004442-1G 004442-5G 004442-25G
FLUO004442-1GEA 0 EUR
FLUO004442-1G FLUO004442-5G FLUO004442-25G
2-Chlor-6-fluorbenzylamin
2-Chlor-6-fluorbenzylamin
Formel: C₇H₇ClFN
Molecular Weight: 159.59 g/mol
Siedepunkt: 99…100 °C (1013 hPa)
MDL: MFCD00042458
CAS-Nummer: 15205-15-9
EINECS: 239-259-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR