1-Chlor-5-fluor-2-iodbenzol
Lieferant: FLUOROCHEM
Synonyme:
2-Chlor-4-fluor-1-iodbenzol
004452-25G
004452-100G
004452-10G
FLUO004452-25GEA
66.8
EUR
FLUO004452-25G
FLUO004452-100G
FLUO004452-10G
1-Chlor-5-fluor-2-iodbenzol
1-Chlor-5-fluor-2-iodbenzol
Formel:
C₆H₃ClFI Molecular Weight: 256.45 g/mol |
MDL:
MFCD00051774 CAS-Nummer: 101335-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...