N-Hydroxysuccinimid

Lieferant: FLUOROCHEM

Synonyme: 1-Hydroxyazacyclopentan-2,5-dion, 1-Hydroxy-1,4-butandisäureimid, NHS, 1-Hydroxybernsteinsäureimid, 1-Hydroxy-2,5-pyrrolidindion, HOSu, N-Hydroxybernsteinsäureimid, N-Hydroxy-1,4-butandisäureimid

005022-100G 005022-250G
FLUO005022-100GEA 60.9 EUR
FLUO005022-100G FLUO005022-250G
N-Hydroxysuccinimid
N-Hydroxysuccinimid
Formel: C₄H₅NO₃
Molecular Weight: 115.09 g/mol
Siedepunkt: 363
Schmelzpunkt: 93…95 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005516
CAS-Nummer: 6066-82-6
EINECS: 228-001-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR