3-Methylfuran
Lieferant: FLUOROCHEM
005088-1G
005088-5G
FLUO005088-1GEA
296
EUR
FLUO005088-1G
FLUO005088-5G
3-Methylfuran
3-Methylfuran
Formel:
C₅H₆O Molecular Weight: 82.1 g/mol Siedepunkt: 65…66 °C (1013 hPa) Dichte: 0.923 g/cm³ (18 °C) |
MDL:
MFCD00060134 CAS-Nummer: 930-27-8 UN: 1992 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...