2'-Chlor-6'-fluoracetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(2-Chloro-6-fluorophenyl)ethanone

005610-5G 005610-1G
FLUO005610-5GEA 82.1 EUR
FLUO005610-5G FLUO005610-1G
2'-Chlor-6'-fluoracetophenon
2'-Chlor-6'-fluoracetophenon
Formel: C₈H₆ClFO
Molecular Weight: 172.59 g/mol
MDL: MFCD01631390
CAS-Nummer: 87327-69-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR