α-Brom-2-chlor-4-fluortoluol

Lieferant: FLUOROCHEM

Synonyme: 2-Chlor-4-fluorbenzylbromid

005630-1G 005630-50G 005630-10G
FLUO005630-1GEA 31.1 EUR
FLUO005630-1G FLUO005630-50G FLUO005630-10G
α-Brom-2-chlor-4-fluortoluol
α-Brom-2-chlor-4-fluortoluol
Formel: C₇H₅BrClF
Molecular Weight: 223.47 g/mol
Schmelzpunkt: 33…35 °C
MDL: MFCD00236025
CAS-Nummer: 45767-66-6
UN: 3261
ADR: 8 II

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR