4-Chlor-3-(trifluormethyl)benzylbromid

Lieferant: FLUOROCHEM

Synonyme: 1-(Brommethyl)-4-chlor-3-(trifluormethyl)benzol

005719.1G 005719-5G 005719-25G
FLUO005719.1GEA 24.2 EUR
FLUO005719.1G FLUO005719-5G FLUO005719-25G
4-Chlor-3-(trifluormethyl)benzylbromid
4-Chlor-3-(trifluormethyl)benzylbromid
Formel: C₈H₅BrClF₃
Molecular Weight: 273.48 g/mol
MDL: MFCD01631540
CAS-Nummer: 261763-23-9
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR