2,6-Difluor-3-methylbenzylbromid

Lieferant: FLUOROCHEM

005759-5G 005759-1G
FLUO005759-5GEA 243 EUR
FLUO005759-5G FLUO005759-1G
2,6-Difluor-3-methylbenzylbromid
2,6-Difluor-3-methylbenzylbromid
Formel: C₈H₇BrF₂
MDL: MFCD01631336
CAS-Nummer: 261763-44-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR