3-Fluor-4-methylbenzylbromid

Lieferant: FLUOROCHEM

Synonyme: 4-(Bromomethyl)-2-fluoro-1-methylbenzene

005799-1G 005799-5G
FLUO005799-1GEA 77.1 EUR
FLUO005799-1G FLUO005799-5G
3-Fluor-4-methylbenzylbromid
3-Fluor-4-methylbenzylbromid
Formel: C₈H₈BrF
Molecular Weight: 203.05 g/mol
MDL: MFCD01631534
CAS-Nummer: 145075-44-1
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR