4-Fluor-3-methylbenzylbromid
Lieferant: FLUOROCHEM
Synonyme:
4-(Bromomethyl)-1-fluoro-2-methylbenzene
005800-1G
005800-5G
005800-10G
FLUO005800-1GEA
69.8
EUR
FLUO005800-1G
FLUO005800-5G
FLUO005800-10G
4-Fluor-3-methylbenzylbromid
4-Fluor-3-methylbenzylbromid
Formel:
C₈H₈BrF |
MDL:
MFCD01631425 CAS-Nummer: 261951-70-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...