5-Fluor-2-methylbenzylbromid

Lieferant: FLUOROCHEM

Synonyme: 2-(Bromomethyl)-4-fluoro-1-methylbenzene

005801-1G 005801-5G 005801-10G
FLUO005801-1GEA 51.9 EUR
FLUO005801-1G FLUO005801-5G FLUO005801-10G
5-Fluor-2-methylbenzylbromid
5-Fluor-2-methylbenzylbromid
Formel: C₈H₈BrF
Molecular Weight: 203.05 g/mol
MDL: MFCD01631528
CAS-Nummer: 261951-71-7
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR