4-Methoxy-3-(trifluormethyl)benzoesäure

Lieferant: FLUOROCHEM

005828-5G 005828-10G
FLUO005828-5GEA 67.1 EUR
FLUO005828-5G FLUO005828-10G
4-Methoxy-3-(trifluormethyl)benzoesäure
4-Methoxy-3-(trifluoromethyl)benzoic acid
Formel: C₉H₇F₃O₃
Molecular Weight: 220.15 g/mol
MDL: MFCD01091014
CAS-Nummer: 213598-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR