5-Bromnicotinonitril

Lieferant: FLUOROCHEM

Synonyme: 3-Brom-5-pyridincarbonitril, 5-Bromnicotinsäurenitril, 3-Brom-5-pyridincarbonsäurenitril, 3-Brom-5-cyanpyridin

CFF361.2.5 CFF361.10 CFF361.50 005886-5G 005886-25G 005886-1G
CHMPCFF361.2.5EA 268 EUR
CHMPCFF361.2.5 CHMPCFF361.10 CHMPCFF361.50 FLUO005886-5G FLUO005886-25G FLUO005886-1G
5-Bromnicotinonitril
5-Bromnicotinonitril
Formel: C₆H₃BrN₂
Molecular Weight: 183,01 g/mol
MDL: MFCD00174363
CAS-Nummer: 35590-37-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR