3,5-Bis(trifluormethyl)benzylamin

Lieferant: FLUOROCHEM

Synonyme: (3,5-Bis(trifluoromethyl)phenyl)methanamine

006021-25G 006021-5G
FLUO006021-25GEA 72.9 EUR
FLUO006021-25G FLUO006021-5G
3,5-Bis(trifluormethyl)benzylamin
3,5-Bis(trifluormethyl)benzylamin
Formel: C₉H₇F₆N
Molecular Weight: 243.15 g/mol
Siedepunkt: 82…84 °C (15 mmHg)
Schmelzpunkt: 46…55 °C
MDL: MFCD00009909
CAS-Nummer: 85068-29-7
EINECS: 285-290-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR