2,4-Bis(trifluormethyl)benzylbromid

Lieferant: FLUOROCHEM

Synonyme: 1-(Brommethyl)-2,4-bis(trifluormethyl)benzol

006022-5G 006022-1G
FLUO006022-5GEA 434 EUR
FLUO006022-5G FLUO006022-1G
2,4-Bis(trifluormethyl)benzylbromid
2,4-Bis(trifluormethyl)benzylbromid
Formel: C₉H₅BrF₆
Molecular Weight: 307.03 g/mol
Siedepunkt: 112 °C (0.2 mmHg)
Dichte: 1.637 g/cm³ (20 °C)
MDL: MFCD00010306
CAS-Nummer: 140690-56-8
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR