3-Fluorbenzylisothiocyanat

Lieferant: FLUOROCHEM

006203-1G 006203-5G
FLUO006203-1GEA 70.2 EUR
FLUO006203-1G FLUO006203-5G
3-Fluorbenzylisothiocyanat
3-Fluorbenzylisothiocyanat
Formel: C₈H₆FNS
Molecular Weight: 167.21 g/mol
MDL: MFCD00060406
CAS-Nummer: 63351-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR