4-Fluorbenzylisothiocyanat
Lieferant: FLUOROCHEM
Synonyme:
1-Fluor-4-(isothiocyanatomethyl)benzol
006204-5G
006204-25G
FLUO006204-5GEA
69.8
EUR
FLUO006204-5G
FLUO006204-25G
4-Fluorbenzylisothiocyanat
4-Fluorbenzylisothiocyanat
Formel:
C₈H₆FNS Molecular Weight: 167.21 g/mol Siedepunkt: 90 °C (0.4 mmHg) Dichte: 1.22 g/cm³ (20 °C) |
MDL:
MFCD00041115 CAS-Nummer: 2740-88-7 EINECS: 220-368-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...