4-Fluorbenzylmercaptan

Lieferant: FLUOROCHEM

Synonyme: p-Fluortoluol-α-thiol

006206-25G 006206-5G
FLUO006206-25GEA 0 EUR
FLUO006206-25G FLUO006206-5G
4-Fluorbenzylmercaptan
4-Fluorbenzylmercaptan
Formel: C₇H₇FS
Molecular Weight: 142.2 g/mol
Siedepunkt: 37 °C (0.1 mmHg)
Dichte: 1.157 g/cm³ (20 °C)
MDL: MFCD00040724
CAS-Nummer: 15894-04-9
EINECS: 240-031-9
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR