2-Fluor-p-anisaldehyd

Lieferant: FLUOROCHEM

Synonyme: 2-Fluor-4-anisaldehyd, 2-Fluor-4-methoxybenzaldehyd

006263-5G 006263-25G
FLUO006263-5GEA 57.9 EUR
FLUO006263-5G FLUO006263-25G
2-Fluor-p-anisaldehyd
2-Fluor-p-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154.14 g/mol
MDL: MFCD00236679
CAS-Nummer: 331-64-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR