5-Fluor-o-toluonitril

Lieferant: FLUOROCHEM

006279-1G 006279-5G
FLUO006279-1GEA 31.1 EUR
FLUO006279-1G FLUO006279-5G
5-Fluor-o-toluonitril
5-Fluor-o-toluonitril
Formel: C₈H₆FN
Molecular Weight: 135,14 g/mol
Siedepunkt: 226 °C (1013 hPa)
Schmelzpunkt: 42…46 °C
MDL: MFCD00042295
CAS-Nummer: 77532-79-7
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR