4-Fluor-m-kresol

Lieferant: FLUOROCHEM
Achtung

Synonyme: 4-Fluor-3-methylphenol

006284-5G 006284-25G
FLUO006284-5GEA 31.1 EUR
FLUO006284-5G FLUO006284-25G
4-Fluor-m-kresol
4-Fluor-m-kresol
Formel: C₇H₇FO
Molecular Weight: 126,13 g/mol
MDL: MFCD00042296
CAS-Nummer: 452-70-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR