2-Fluorphenetol

Lieferant: FLUOROCHEM

Synonyme: 1-Fluor-2-ethoxybenzol, (2-Fluorphenyl)ethylether

006316-1G 006316-5G
FLUO006316-1GEA 36.9 EUR
FLUO006316-1G FLUO006316-5G
2-Fluorphenetol
2-Fluorphenetol
Formel: C₈H₉FO
Molecular Weight: 140,16 g/mol
MDL: MFCD00017909
CAS-Nummer: 451-80-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR