1-(4-Fluorphenyl)pyrrol
Lieferant: FLUOROCHEM
Synonyme:
N-(4-Fluorphenyl)pyrrol
006341-1G
006341-10G
FLUO006341-1GEA
0
EUR
FLUO006341-1G
FLUO006341-10G
1-(4-Fluorphenyl)pyrrol
1-(4-Fluorophenyl)pyrrole
Formel:
C₁₀H₈FN Molecular Weight: 161,18 g/mol |
MDL:
MFCD00047066 CAS-Nummer: 81329-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...