2-Fluor-3-(trifluormethyl)benzaldehyd

Lieferant: FLUOROCHEM

006380-1G 006380-5G 006380-10G
FLUO006380-1GEA 31.1 EUR
FLUO006380-1G FLUO006380-5G FLUO006380-10G
2-Fluor-3-(trifluormethyl)benzaldehyd
2-Fluoro-3-(trifluoromethyl)benzaldehyde
Formel: C₈H₄F₄O
Molecular Weight: 192,11 g/mol
MDL: MFCD00061246
CAS-Nummer: 112641-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR