2-Fluor-4-(trifluormethyl)benzaldehyd

Lieferant: FLUOROCHEM

006381-1G 006381-5G 006381-10G
FLUO006381-1GEA 31.1 EUR
FLUO006381-1G FLUO006381-5G FLUO006381-10G
2-Fluor-4-(trifluormethyl)benzaldehyd
2-Fluoro-4-(trifluoromethyl)benzaldehyde
Formel: C₈H₄F₄O
Molecular Weight: 192,11 g/mol
Siedepunkt: 118…119 °C (1013 hPa)
Dichte: 1,41 g/cm³ (20 °C)
MDL: MFCD00061310
CAS-Nummer: 89763-93-9
UN: 1987
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR