2-Fluor-5-(trifluormethyl)benzaldehyd
Lieferant: FLUOROCHEM
006382-1G
006382-5G
006382-25G
FLUO006382-1GEA
31.1
EUR
FLUO006382-1G
FLUO006382-5G
FLUO006382-25G
2-Fluor-5-(trifluormethyl)benzaldehyd
2-Fluoro-5-(trifluoromethyl)benzaldehyde
Formel:
C₈H₄F₄O Molecular Weight: 192,11 g/mol |
MDL:
MFCD00061311 CAS-Nummer: 146137-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...