3-Fluor-5-(trifluormethyl)benzaldehyd

Lieferant: FLUOROCHEM

006384-1G 006384-5G 006384-10G
FLUO006384-1GEA 31.1 EUR
FLUO006384-1G FLUO006384-5G FLUO006384-10G
3-Fluor-5-(trifluormethyl)benzaldehyd
3-Fluoro-5-(trifluoromethyl)benzaldehyde
Formel: C₈H₄F₄O
MDL: MFCD00061312
CAS-Nummer: 188815-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR