4-Fluor-3-(trifluormethyl)benzaldehyd

Lieferant: FLUOROCHEM

006386-1G 006386-5G 006386-10G
FLUO006386-1GEA 0 EUR
FLUO006386-1G FLUO006386-5G FLUO006386-10G
4-Fluor-3-(trifluormethyl)benzaldehyd
4-Fluoro-3-(trifluoromethyl)benzaldehyde
Formel: C₈H₄F₄O
MDL: MFCD00061314
CAS-Nummer: 67515-60-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR