3-Fluor-5-(trifluormethyl)benzylamin

Lieferant: FLUOROCHEM

Synonyme: (3-Fluoro-5-(trifluoromethyl)phenyl)methanamine

006419-1G 006419-5G
FLUO006419-1GEA 101 EUR
FLUO006419-1G FLUO006419-5G
3-Fluor-5-(trifluormethyl)benzylamin
3-Fluor-5-(trifluormethyl)benzylamin
Formel: C₈H₇F₄N
Molecular Weight: 193.14 g/mol
Siedepunkt: 185
Schmelzpunkt: 46
Dichte: 1.319
MDL: MFCD00061163
CAS-Nummer: 150517-77-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR